missing translation for 'onlineSavingsMsg'
Learn More

LAP (TGF-beta 1) Antibody (27235) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30733233 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733233 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733233 Supplier Novus Biologicals Supplier No. FAB246JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Neutralization,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP (TGF-beta 1)
Applications Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 27235
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-β1)
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.