missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET VDR Coactivator Assay Kit, goat

Product Code. 10138962
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10138962 800 x 20 μL assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10138962 Supplier Thermo Scientific™ Supplier No. PV4554

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Contains Goat Tb-Anti-GST antibody

  • This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15127
  • The LanthaScreen™ TR-FRET Vitamin D Receptor (VDR) Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential VDR ligands as agonists of coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a VDR ligand-binding domain (VDR-LBD) that is tagged with glutathione-S-transferase (GST), in a homogenous mix-and-read assay format
  • To run the LanthaScreen™ TR-FRET VDR Coactivator Assay, VDR-LBD is added to ligand test compounds, followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-anti-GST antibody. After room temperature incubation, the TR-FRET 520:495 emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound.
  • Based upon the biology of the VDRcoactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).
  • The LanthaScreen™ TR-FRET VDR Coactivator Assay Kit contains the VDR-LBD (GST) protein, fluorescein-labeled TRAP220/DRIP-2 coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Quantity 800 x 20 μL assays
Packaging 384-well Plate
Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Assay Entry Biochemical coregulator interaction
Gene ID (Entrez) 7421
Readout End Point
Shipping Condition Dry Ice
Target Entry VDR, NR1I1
Product Line LanthaScreen
For Use With (Application) Co-Factor Interaction Assay, TR-FRET
Ligand VDR
Product Type TR-FRET VDR Coactivator Assay Kit
No. of Assays 800 x 20 μL Assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.