missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET TR alpha Coactivator Assay Kit, goat

Product Code. 10276503
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10276503

Brand: Thermo Scientific™ PV4687

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The LanthaScreen™ TR-FRET TR (thyroid receptor) alpha Coactivator Assay Kit provides a sensitive and robust method for highthroughput screening of potential TR alpha ligands as agonists of ligand-dependent coactivator recruitment.

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15136:

The LanthaScreen™ TR-FRET TR (thyroid receptor) alpha Coactivator Assay Kit provides a sensitive and robust method for highthroughput screening of potential TR alpha ligands as agonists of ligand-dependent coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human recombinant TR alpha ligand-binding domain (TR alpha-LBD) that is tagged with glutathione-S-transferase (GST) in a homogeneous, mix-and-read assay format.

To Assay:

When performing the LanthaScreen™ TR-FRET TR alpha coactivator assay, TR alpha-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and used to determine the EC50 from a dose response curve of the compound. Based on the biology of the TR alpha-coactivator peptide interaction, the ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, induce a conformational change, and recruit coactivator peptide (Figure 1).

Contents and Storage:

The LanthaScreen™ TR-FRET TR alpha Coactivator Assay Kit contains TR alpha-LBD (GST) protein, fluorescently labeled SRC2-2 coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)
Description LanthaScreen™ TR-FRET TR Alpha Coactivator Assay Kit, 800 x 20μL Assays, 384-Well Plate Format, Tb (Terbium) Dye Type, Fluorescent Detection Method, TR-FRET Technique, For Pharma & Biopharma, Target & Lead Identification & Validation
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Includes TR alpha-LBD (GST) protein, fluorescently labeled SRC2-2 coactivator peptide, Tb-labeled anti-GST antibody, and buffers
Quantity 800 x 20 μL assays
Regulatory Status For research use only (RUO)
Packaging 384-well Plate
Content And Storage Store in ultra-cold freezer (-68°C to -85°C).
Assay Entry Biochemical coregulator interaction
Readout End Point
Shipping Condition Dry Ice
Target Entry THRA, TR alpha, NR1A1
Product Line LanthaScreen™
For Use With (Application) Co-Factor Interaction Assay, TR-FRET
Ligand TR alpha
Product Type TR-FRET TR Alpha Coactivator Assay Kit
No. of Assays 800 x 20 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.