missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET PPAR alpha Coactivator Assay Kit, rabbit

Product Code. 15343796
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15343796

Brand: Thermo Scientific™ A15133

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The LanthaScreen™ TR-FRET PPAR (peroxisome proliferator activated receptor) alpha Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential PPAR alpha ligands as agonists or antagonists of ligand-dependent coactivator recruitment.

This kit contains Rabbit Tb-Anti-GST antibody; other kit components are the same as kit PV4684:

The LanthaScreen™ TR-FRET PPAR (peroxisome proliferator activated receptor) alpha Coactivator Assay provides a sensitive and robust method for high-throughput screening of potential PPAR alpha ligands as agonists or antagonists of ligand-dependent coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human recombinant PPAR alpha ligand-binding domain (PPAR alpha-LBD) that is tagged with glutathione-S-transferase (GST) in a homogeneous mix-and-read assay format.

Agonist mode:

When running the LanthaScreen™ TR-FRET PPAR alpha Coactivator Assay in agonist mode (to identify agonist compounds), PPAR alpha- LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the PPAR alpha-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (see Figure 1).

Antagonist mode:

When the LanthaScreen™ TR-FRET PPAR alpha Coactivator Assay is run in antagonist mode (to identify antagonist compounds), PPAR alpha-LBD is added to ligand test compounds followed by addition of a mixture of agonist, fluorescein-coactivator peptide, and Tb-labeled anti-GST antibody (Figure 2). The concentration of agonist used in this mode is the EC80 concentration as determined by first running the assay in agonist mode.

Contents and Storage:

The LanthaScreen™ TR-FRET PPAR alpha Coactivator Assay Kit contains PPAR alpha-LBD (GST) protein, fluorescently labeled PGC1a coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or 4°C).

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Conjugate Tb (Terbium)
Description LanthaScreen™ TR-FRET PPAR Alpha Coactivator Assay Kit, 800 x 20μL Assays, Green, 384-Well Plate Format, Tb (Terbium) Dye Type, Fluorescent Detection Method, TR-FRET Technique, For Pharma & Biopharma, Target Lead Identification & Validation
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Includes Tb-anti-GST antibody (rabbit), PPAR alpha-LBD, GST, fluorescein coactivator peptide, TR-FRET coregulator buffer J, 1M DTT
Quantity 800 x 20 μL assays
Regulatory Status For research use only (RUO)
Packaging 384-well Plate
Content And Storage • Tb-anti-GST Antibody (Rabbit) (store at -20°C)
• PPAR alpha-LBD, GST (-80°C)
• Fluoroscein Coactivator Peptide (-20°C)
• TR-FRET Coregulatore Buffer J (4°C)
• 1M DTT (-20°C or -80°C)
Assay Category Biochemical Assay
Druggable Target Nuclear Receptors
Readout TR-FRET (Activity)
Product Line LanthaScreen
For Use With (Application) Nuclear Receptor Assay, TR-FRET
Ligand PPAR alpha
Product Type TR-FRET PPAR Alpha Coactivator Assay Kit
No. of Assays 800 x 20 μL Assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.