missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ TR-FRET FXR Coactivator Assay Kit, goat
Description
This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15140:
The LanthaScreen™ TR-FRET FXR (farnesoid X receptor) Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential FXR ligands as agonists of coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein- labeled coactivator peptide, and an FXR ligand-binding domain (FXR-LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous, mix-and-read assay format.
To assay:
When running the LanthaScreen™ TR-FRET FXR coactivator assay in agonist mode, FXR-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and used to determine the EC50 from a dose response curve of the compound. Based on the biology of the FXR-coactivator peptide interaction, the ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).
The LanthaScreen™ TR-FRET FXR (farnesoid X receptor) Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential FXR ligands as agonists of coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein- labeled coactivator peptide, and an FXR ligand-binding domain (FXR-LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous, mix-and-read assay format.
To assay:
When running the LanthaScreen™ TR-FRET FXR coactivator assay in agonist mode, FXR-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and used to determine the EC50 from a dose response curve of the compound. Based on the biology of the FXR-coactivator peptide interaction, the ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).
Specifications
Specifications
| Conjugate | Tb (Terbium) |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Microplate Reader |
| Quantity | 800 x 20 μL assays |
| Packaging | 384-well Plate |
| Content And Storage | The LanthaScreen™ TR-FRET FXR Coactivator Assay Kit contains FXR-LBD (GST) protein, fluorescently labeled SRC2-2 coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C). |
| Assay Entry | Biochemical coregulator interaction |
| Gene ID (Entrez) | 9971 |
| Readout | End Point |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?