missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ TR-FRET CAR Coactivator Assay Kit, goat

Product Code. 10428853
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10428853 800 x 20 μL assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10428853 Supplier Thermo Scientific™ Supplier No. PV4836

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The LanthaScreen™ TR-FRET Constitutive Androstane Receptor (CAR) Coactivator Assay provides a sensitive and robust method for highthroughput screening (HTS) of potential CAR ligands as agonists of ligand-dependent coactivator recruitment or inverse agonists of ligand-dependent coactivator displacement.

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15141:

The LanthaScreen™ TR-FRET Constitutive Androstane Receptor (CAR) Coactivator Assay provides a sensitive and robust method for highthroughput screening (HTS) of potential CAR ligands as agonists of ligand-dependent coactivator recruitment or inverse agonists of ligand-dependent coactivator displacement. The kit uses a terbium-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a CAR ligand-binding domain (CAR-LBD) that is tagged with glutathione-S-transferase (GST) in a homogeneous mix-and-read assay format.

To assay:

When running the LanthaScreen™ TR-FRET CAR Coactivator Assay, CAR-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and terbium anti-GST antibody. After an incubation period at room temperature, the 520 nm/ 495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the CAR-coactivator peptide interaction, this ligand's EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and either recruit or displace coactivator peptide (Figures 1 and 2).

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Tb (Terbium)
Description LanthaScreen™ TR-FRET CAR Coactivator Assay Kit, 800 x 20μL Assays, 384-Well Plate Format, Tb (Terbium) Dye Type, Fluorescent Detection Method, TR-FRET Technique, For Pharma & Biopharma, Target & Lead Identification & Validation
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Includes CAR-LBD (GST) protein, fluorescently labeled PGC1a coactivator peptide, terbium-labeled anti-GST antibody, and buffers
Quantity 800 x 20 μL assays
Regulatory Status For research use only (RUO)
Packaging 384-well Plate
Content And Storage The LanthaScreen™ TR-FRET CAR Coactivator Assay Kit contains CAR-LBD (GST) protein, fluorescently labeled PGC1a coactivator peptide, terbium-labeled anti-GST antibody, and buffers.

Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
Assay Entry Biochemical coregulator interaction
Readout End Point
Shipping Condition Dry Ice
Target Entry CAR, NR1I3
Product Line LanthaScreen
For Use With (Application) Co-Factor Interaction Assay, TR-FRET
Ligand CAR
Product Type TR-FRET CAR Coactivator Assay Kit
No. of Assays 800 x 20 μL Assays
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.