missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-pMAP2K1 (pSer217/221) Antibody Kit
Description
A member of the MAP Kinase family, MAP2K1 (mitogen-activated protein kinase kinase 1) is phosphorylated at Ser217/221 by a range of MAPK family members including Raf and B-Raf. For use in combination with the Fluorescein-MAP2K1 physiological protein substrate (PV4812) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Applications | Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay |
| Conjugate | Tb (Terbium)/FITC/Lanthanide Chelates |
| Gene Alias | COT,EST,ESTF,FLJ10486,MEKK8,TPL2,Tpl-2,c-CO, CRAF,NS5,Raf-1,c-Ra, MEKK9,MLK1,PRKE, COT,EST,ESTF,FLJ10486,MEKK8,TPL2,Tpl-2,c-CO, MEKK7,TAK1,TGF1, MEKK9,MLK1,PRKE, ASK1,MAPKKK5,MEKK, MEKK10,MLK2,MS, MEKK11,MGC17114,MLK-3,MLK3,PTK1,SPR, CRAF,NS5,Raf-1,c-Ra, |
| Molecular Weight of Antigen | 150kDa |
| Quantity | 25 μg |
| Regulatory Status | RUO |
| Research Discipline | Target and Lead Identification and Validation |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 10454, 10746, 1326, 4215, 4217, 4293, 4294, 4296, 5894, 673, 6885 |
| Packaging | 1 Kit(s) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?