Learn More
Thermo Scientific™ LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit
LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit
Brand:  Thermo Scientific™ PV4451
 View more versions of this product
Product Code. 11525046
  • £3805.00 / 25µg
For Research Use Only. All usage must comply with product instructions.
Description
Description
A member of the leucine zipper family of DNA binding proteins, ATF2 (activating transcription factor 2) is phosphorylated at Ser71 by members of the JNK and p38 families of kinases.
For use with the GFP-ATF2 physiological protein substrate (PV4445) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serine/threonine kinase
Tb (Terbium)/GFP (EmGFP)
384-well plate
RUO
Primary
Terbium-labeled anti-ATF2 [pSer71] antibody in HEPES buffered saline. Store at -20°C.
Biochemical terbium phospho-specific activity
340⁄490, 520
Dry Ice
Hu
Serine⁄Threonine Kinases
MAPK1, MAPK3, MAPK8, MAPK9, MAPK10, MAPK11, MAPK12, MAPK13, MAPK14
Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay
LanthaScreen™ Tb-pATF2 Antibody Kit, 25μg, 150kDa Molecular Weight, TR-FRET Technique, Ser/Thr kinases Antigen, Human Species Reactivity, GFP (EmGFP), Tb (Terbium) Conjugate, Used For Biochemical Kinase Assays, Drug Discovery and Development
25 μg
Protein Biology
1 Kit(s)
150000 Da
384-well Plate
Human
End Point
MAPK13, MAPK8, MAPK9, MAPK14, MAPK3, MAPK1, MAPK11, MAPK12, MAPK10
TR-FRET
LanthaScreen