Learn More
Thermo Scientific™ LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit
LanthaScreen™ Tb-pATF2 (pSer71) Antibody Kit
£3805.00 - £16330.00
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11525046
View Documents
Thermo Scientific™
PV4451
25 μg
£3805.00
25µg
View alternative products
Save Up to 
»
10175772
View Documents
Thermo Scientific™
PV4452
1 mg
£16330.00
1mg
View alternative products
Save Up to 
»
Description
Description
A member of the leucine zipper family of DNA binding proteins, ATF2 (activating transcription factor 2) is phosphorylated at Ser71 by members of the JNK and p38 families of kinases.
For use with the GFP-ATF2 physiological protein substrate (PV4445) to evaluate inhibitors in medium to high throughput screening applications.