Learn More
Thermo Scientific™ LanthaScreen™ Tb-CREB [pSER133] Antibody
A member of the leucine zipper family of DNA binding proteins, CREB (cAMP responsive element binding) protein is phosphorylated at Ser133 by a wide range of Ser/Thr protein kinases.
Brand: Thermo Scientific™ PV3542
Description
A member of the leucine zipper family of DNA binding proteins, CREB (cAMP responsive element binding) protein is phosphorylated at Ser133 by a wide range of Ser/Thr protein kinases.
For use with the Fluorescein-CREBtide peptide substrate (Cat. No. PV3508) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
| Serine/threonine kinase | |
| 1.1mg/mL | |
| 25 μg | |
| Pharma and Biopharma | |
| 1 Each | |
| Liquid | |
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| AMPK (A2/B1/G1), STK22D, CAMK2B, CHEK1, MAPKAPK2, PASK, PIM2, PRKCN, PRKD2, PRKG2, RPS6KA5, AMPK (A1/B1/G1), AURKA, CAMK1D, CAMK4, CHEK2, MAPKAPK3, PIM1, PKN1, PRKD1, PRKG1, RPS6KA4, CAMK1 |
| Kinase Assay | |
| Tb (Terbium)/Lanthanide Chelates | |
| RUO | |
| Primary | |
| Terbium-labeled anti-phosphoserine peptide-specific antibody in 10 mM HEPES (pH 7.5), 137 mM NaCl, and 2.7 mM KCl (HEPES buffered saline) | |
| Biochemical terbium phospho-specific activity | |
| Human | |
| End Point | |
| Ser/Thr Kinases | |
| LanthaScreen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.