Learn More
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody
Brand:  Invitrogen PV5129
Code : 50
Additional Details : Weight : 0.00900kg
 View more versions of this product
Product Code. 10083614
  • £17700.00 / Each
Estimated Shipment: 10-05-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
The p53 tumor suppressor protein is an essential component of many different signaling pathways. The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR.
For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (PV5132) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
Ser/Thr kinases
Tb (Terbium)
1 mg
Pharma and Biopharma
1 Each
Phosphorylation of p53
Monoclonal
150kDa
RUO, RUO
Primary
–20°C