missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody
Description
- The LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody is a key component of the BacMam Histone H3K4me3 Cellular Assay (A14163)
- The LanthaScreen™ Histone H3K4me3 antibody allows investigation of trimethylation at lysine 4 on histone H3
- This enables screening for potential inhibitors of methyl transferases and demethylases associated with Lys4 trimethylation in their natural complexes
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K4me3 Antibody Lets You:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Specifications
Specifications
| Concentration | 3500 nM, 0.5 mg/mL |
| Conjugate | Tb (Terbium)/Lanthanide Chelates |
| Quantity | 100 μg |
| Content And Storage | Store the LanthaScreen™ Tb-anti-histone H3K4me3 Antibody at -20°C. |
| Form | Liquid |
| Shipping Condition | Dry Ice |
| Target Modification State | Methylated |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?