missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K4me2 Antibody

The LanthaScreen™ Tb-anti-Histone H3K4me2 antibody is a key component of the BacMam Histone H3K4me2 Cellullar Assay (A14164).

£6720.00 - £31670.00

Specifications

Conjugate Tb (Terbium)/Lanthanide Chelates
Concentration 3500 nM, 0.5 mg/mL
Form Liquid
Shipping Condition Dry Ice
Target Modification State Methylated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11692049
View Documents
Thermo Scientific™
A14158
100 μg
£6720.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11662049
View Documents
Thermo Scientific™
A14153
1 mg
£31670.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The LanthaScreen™ Tb-anti-Histone H3K4me2 antibody is a key component of the BacMam Histone H3K4me2 Cellullar Assay (A14164)
  • When this terbium (Tb)- labeled antibody is paired with baculovirus-mediated (BacMam) expression of a Histone H3-GFP fusion protein, the TR-FRET readout enables sensitive detection of H3 lysine 4 dimethylation
  • The resulting high-throughput screening (HTS) compatible assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 4 of histone H3 in the cell type of your choice
  • For research use only
  • Not for human or animal therapeutic or diagnostic use

LanthaScreen™ Tb-anti-Histone H3K4me2 Antibody Allows Users to:

  • Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
  • Conserve precious cell samples with the miniaturizable homogenous assay format
  • Improve data quality with the advantages of TR-FRET
Specifications

Specifications

Tb (Terbium)/Lanthanide Chelates
Liquid
Methylated
3500 nM, 0.5 mg/mL
Dry Ice
Videos
SDS
Documents

Documents

Certificates

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.