Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-ERK2 [pThr185/pTyr187] Antibody
A part of the MAPK Pathway, ERK2 is phosphorylated at Thr185 and Tyr187 by the mitogen-activated protein kinase kinases 2 (MAP2K2).
Brand: Thermo Scientific™ PV5210
This item is not returnable.
View return policy
Description
- A part of the MAPK Pathway, ERK2 is phosphorylated at Thr185 and Tyr187 by the mitogen-activated protein kinase kinases 2 (MAP2K2)
- For use in combination with the LanthaScreen™ ERK2 A375 cell line (K1798) to evaluate inhibitors in medium to high throughput screening applications
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled pERK2 (pThr185/pTyr187) antibody in HEPES buffered saline. Store at -20°C. | |
| Human | |
| End Point | |
| Ser/Thr Kinases, Tyrosine Kinases |
| 1 mg | |
| 340/490, 520 | |
| Dry Ice | |
| Hu | |
| LanthaScreen |