missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ PRAS40 293E Cell Line

Product Code. 10105073
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10105073 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10105073 Supplier Thermo Scientific™ Supplier No. K1528

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The PI3K/AKT pathway is activated by various stimuli (including insulin and IGF-1) and mediates signals for cell growth, cell survival, translation, and inhibition of apoptosis.

The PI3K/AKT pathway is activated by various stimuli (including insulin and IGF-1) and mediates signals for cell growth, cell survival, translation, and inhibition of apoptosis. PRAS40 (Proline-rich AKT substrate, 40 kDa) is a cytosolic protein that is phosphorylated at position Thr246 by AKT directly upon stimulation of the pathway. PRAS40 has been shown to bind to the mTORC1 complex and inhibit downstream signaling (i.e., phosphorylation of 4E-BP1 or p70S6K). LanthaScreen™ PRAS40 HEK293E is a human cell line which constitutively expresses GFP-PRAS40 fusion proteins.

The kinase substrate was introduced using lentiviral transduction followed by selection with blasticidin. This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as a sorting marker. The PI3K/AKT pathway is known to be active in the highly insulin-responsive HEK293E cell background. Using this cell line, a homogenous ('addition only') immunoassay has been developed in a 384-well format. The phosphorylation of PRAS40 at Thr246 is detected in cell lysates using a terbium-labeled phosphospecific antibody in a time-resolved FRET (TR-FRET) readout.

The performance of this assay has been tested using numerous experimental variables, including cell plating density, stimulation time, DMSO tolerance, and antibody incubation time. Under optimized conditions, the assay has been further validated and shows correct EC50 and IC50 values, with insulin as the primary agonist and PI-103 as the known inhibitor. Overall, the assay displays excellent statistical data (Z'-factor >0.6), high signal-to-background (response ratio), and is a robust cell-based readout of AKT signaling.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists Insulin
Antagonists IGF-1
Assay Entry Cell-based terbium phospho-specific activity (stable)
Cell State Dividing Cells
Content And Storage LanthaScreen™ PRAS40 HEK293E cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
For Use With (Application) Kinases, Signaling Pathway
Gene Symbol PRAS40
Primary Agonist Insulin
Product Line LanthaScreen
Quantity 1 Vial
Readout End Point
Reporter Gene GFP (EmGFP)
Target Entry PRAS40
Type Dividing Cells
System Type LanthaScreen™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.