missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Eu-Streptavidin Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates. The Eu-streptavidin Kit contains 10μg Eu-Streptavidin, 400μl AlexaFluor™ 647-biotin control peptide, and 100 mL of TR-FRET Dilution Buffer.
Specifications
Specifications
| Concentration | 0.25 mg/mL |
| Conjugate | Alexa Fluor 647, Europium |
| Quantity | 1 Kit |
| Content And Storage | The Eu-Streptavidin Kit contains 10 μg Eu-streptavidin, 400 μl AlexaFluor™ 647-biotin control peptide and 100 mL of TR-FRET Dilution Buffer. Store reagents as indicated on the certificate of analysis. |
| Excitation/Emission | 340/615, 665 |
| Shipping Condition | Dry Ice |
| Form | Liquid |
| Product Line | LanthaScreen™ |
| Buffer | TR-FRET Dilution Buffer |
| Product Type | Streptavidin Conjugate (Fluorescent) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction