missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine, 96%

Product Code. 15478465
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478465 1 g 1g
15488465 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478465 Supplier Thermo Scientific Alfa Aesar Supplier No. H33439.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 137438-23-4 | C11H11ClN2S | 238.733 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137438-23-4
Molecular Formula C11H11ClN2S
Molecular Weight (g/mol) 238.733
MDL Number MFCD02333821
InChI Key ZSDWARZBRRSRLT-UHFFFAOYSA-N
Synonym 4-chloro-7-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydrobenzo b thiopheno 2,3-d pyrimidine, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6-tetraene, rs-4-chloro-7-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydro-1 benzothiolo 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidine, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 , trideca-1 9 ,2,4,6-tetraene, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0?,? trideca-1 9 ,2,4,6-tetraene, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2 7 ,3,5-tetraene
PubChem CID 2790841
IUPAC Name 4-chloro-7-methyl-5,6,7,8-tetrahydro-[1]benzothiolo[2,3-d]pyrimidine
SMILES CC1CCC2=C(C1)SC3=C2C(=NC=N3)Cl

Specifications

Melting Point 109°C to 113°C
Quantity 1 g
Formula Weight 238.73
Percent Purity 96%
Chemical Name or Material 4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.