Learn More
Thermo Scientific™ LanthaScreen™ Eu-anti-DYKDDDDK Antibody Kit
LanthaScreen™ Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag).
Brand: Thermo Scientific™ PV6034
Description
LanthaScreen™ Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity.
Specifications
| 0.25 mg/mL | |
| LanthaScreen™ Eu-anti-DYKDDDDK Antibody Kit, 1 Kit, 0.25mg/mL Concentration, Alexa Fluor™ 647, Europium Conjugate, 340/615nm Excitation, 665nm Emission, HEPES Buffered Saline (137mM NaCl, 2.7mM KCl, 10mM HEPES pH 7.5) Storage Buffer, Used for | |
| 1 Kit | |
| Target and Lead Identification and Validation | |
| 1 Kit(s) | |
| Frozen | |
| Dry Ice |
| Europium/Lanthanide Chelates | |
| Frozen | |
| RUO | |
| Primary | |
| Europium-labeled anti-DYKDDDDK antibody in HEPES buffered saline. Store at -20°C. | |
| 340/615, 665 nm | |
| LanthaScreen™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.