missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum trifluoromethanesulfonate, anhydrous, 99%

Product Code. 11365475
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11365475

Brand: Thermo Scientific Alfa Aesar L19657.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52093-26-2 | C3F9LaO9S3 | 586.095 g/mol

Lanthanum trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes. It is an effective catalyst for the Baylis-Hillman reaction. It also catalyzes Michael additions to methyl vinyl ketone in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lanthanum trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes. It is an effective catalyst for the Baylis-Hillman reaction. It also catalyzes Michael additions to methyl vinyl ketone in water.

Solubility
Soluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with bases. It is hygroscopic in nature. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-26-2
Molecular Formula C3F9LaO9S3
Molecular Weight (g/mol) 586.095
MDL Number MFCD00077560
InChI Key WGJJZRVGLPOKQT-UHFFFAOYSA-K
Synonym lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum
PubChem CID 2733938
IUPAC Name lanthanum(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3]

Specifications

Linear Formula (CF3SO3)3La
Quantity 25 g
UN Number UN3261
Beilstein 9009103
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 586.1
Percent Purity 99%
Chemical Name or Material Lanthanum trifluoromethanesulfonate, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.