missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Standard: La @ 10000 μg/mL in 5% HNO3

Product Code. 30314955
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. Quantity unitSize
30314955 250 mL 250mL
30312841 100 mL 100mL
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 30314955

Brand: VHG Labs VHGTLAN250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lanthanum
CAS 7439-91-0
Analyte or Component Names Lanthanum
Shipping Condition Room Temperature
Molecular Formula La
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym La
InChI Formula InChI=1 S/La
Solution Type Single Solution
SMILES [La]
Element La
IUPAC Name lanthanum
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 138.906
Quantity 250 mL
Formula Weight 138.906
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.