missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Standard: La @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17929299
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
Product Code. Quantity unitSize
17929299 50 mL 50mL
17958379 100 mL 100mL
17968879 250 mL 250mL
17977769 500 mL 500mL
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 17929299

Brand: VHG Labs VHGPLAN50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lanthanum Standard: La @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lanthanum
CAS 7439-91-0
Methods ICP/ICP-MS
Analyte or Component Names Lanthanum
Shipping Condition Room Temperature
Molecular Formula La
Certifications/Compliance ISO 17034
Synonym La
InChI Formula InChI=1S/La
SMILES [La]
IUPAC Name lanthanum
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 138.906
Quantity 50 mL
Formula Weight 138.906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.