missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 15412557
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
7439-91-0
La
138.91
MFCD00011066
FZLIPJUXYLNCLC-UHFFFAOYSA-N
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
23926
CHEBI:33336
lanthanum
[La]
This item is not returnable. View return policy
7439-91-0
La
138.91
MFCD00011066
FZLIPJUXYLNCLC-UHFFFAOYSA-N
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
23926
CHEBI:33336
lanthanum
[La]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Silver
UN Number UN3089
Quantity 25 g
Percent Purity 99.9%
Mesh Size −200 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Lanthanum powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'