missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Oxide, 99.99%, MP Biomedicals

Product Code. 11447855 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447855 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447855 Supplier MP Biomedicals Supplier No. 0521460410
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity grade lanthanum oxide suitable for research applications.

Specifications

CAS 1312-81-8
Quantity 10 g
Molecular Weight (g/mol) 325.82 g/moL
Formula Weight 325.82
Chemical Name or Material Lanthanum Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.