missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Nitrate Solution According to European Pharmacopoeia (EP), Reagecon

Product Code. 11414699 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414699 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414699 Supplier Reagecon Supplier No. 1048001
  • ONLINE EXCLUSIVE PRICE
    £1660.00
    £1626.71 / 1 L
    Save £33.29
    2% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications including use in the identification of reactions of ions and functional groups (2.3.1), and use as a reagent in the performance of tests outlined in several EP monographs.

  • European Pharmacopoeia Reagent, Solution
  • High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications.
  • Applications include use in the identification reactions of ions and functional groups (2.3.1), and use as a reagent in the performance of tests outlined in several EP monographs.
  • Accurate, stable and reproducible.
  • Stability is proven (either opened or unopened) for the shelf life of this product.

Specifications

Packaging HDPE Bottle GL45
Physical Form Liquid
Technique EP Testing
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.