missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum nickel, REacton™, 99.9% (REO)

Product Code. 11356348
Change view
Click to view available options
Quantity:
50 g
5 x 50 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356348 50 g 50g
11346348 5 x 50 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356348 Supplier Thermo Scientific Alfa Aesar Supplier No. 011271.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12196-72-4 | LaNi5 | 432.37 g/mol

Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12196-72-4
Molecular Formula LaNi5
Molecular Weight (g/mol) 432.37
MDL Number MFCD00198177
InChI Key BUCVQDCWULHLJZ-UHFFFAOYSA-N
Synonym lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy
PubChem CID 14367487
IUPAC Name lanthanum;nickel
SMILES [Ni].[Ni].[Ni].[Ni].[Ni].[La]

Specifications

Quantity 50 g
Odor Odorless
Sensitivity Air and moisture sensitive
Form Pieces
UN Number UN3178
Solubility Information Reacts with water at 0°C.
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Lanthanum nickel
compliance-icons compliance-icons
Produktidentifikator
  • Lanthanum, compound with nickel (1:5)
Signalwort
  • Danger
Gefahrenkategorie
  • Carcinogenicity Category 2
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 1
Gefahrenhinweis
  • H228-Flammable solid.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Ergänzende Informationen
  • MIXTURE LIST-Contain: Lanthanum, compoundwith nickel

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.