missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum nickel powder, -100 mesh, 99.5% (metals basis)

Product Code. 11359389
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359389 25 g 25g
11369389 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11359389 Supplier Thermo Scientific Alfa Aesar Supplier No. 089133.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 12196-72-4 | LaNi5 | 432.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12196-72-4
Molecular Formula LaNi5
Molecular Weight (g/mol) 432.37
MDL Number MFCD00198177
InChI Key BUCVQDCWULHLJZ-UHFFFAOYSA-N
Synonym lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy
PubChem CID 14367487
IUPAC Name lanthanum;nickel
SMILES [Ni].[Ni].[Ni].[Ni].[Ni].[La]

Specifications

Quantity 25 g
Mesh Size −100 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
UN Number UN3178
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Lanthanum nickel powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.