missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) sulfide, 99% (REO)

Product Code. 11378077
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378077 2 g 2g
11388077 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378077 Supplier Thermo Scientific Alfa Aesar Supplier No. 022655.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12031-49-1 | La2S3 | 373.99 g/mol

Lanthanum(III) sulfide is used as a precursor in the production of gallium lanthanum sulfide and strontium lathanum sulfide. It is also used in glass manufacturing. It is doped with other metals and used in optical fibers. It is also used as optical switching, high power-delivery, acousto-optic devices, air-guiding fibers, fiber sensors and mid-IR devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-49-1
Molecular Formula La2S3
Molecular Weight (g/mol) 373.99
MDL Number MFCD00016168
InChI Key YTYSNXOWNOTGMY-UHFFFAOYSA-N
Synonym lanthanum sulfide, dilanthanum trisulfide, dilanthanum trisulphide, lanthanum sesquisulfide, lanthanum 3+ sulfide, lanthanum sulfide la10s15, lanthanum disulfide, lanthanum 3+ trisulfide
PubChem CID 165996
IUPAC Name lanthanum(3+);trisulfide
SMILES [S--].[S--].[S--].[La+3].[La+3]

Specifications

Melting Point 2100° (vacuum)
Color Pale Yellow
Physical Form Powder
Quantity 2 g
Assay Percent Range 99% (REO)
Formula Weight 374.02
Odor Like rotten eggs
Density 4.911
Chemical Name or Material Lanthanum(III) sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.