missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) sulfate hydrate, 99.9% (REO)

Product Code. 11384768
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384768 500 g 500g
11374768 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384768 Supplier Thermo Scientific Alfa Aesar Supplier No. 043106.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57804-25-8 | La2O12S3 | 565.98 g/mol

Used as colorant or laboratory reagent. It is also mainly applied in specialty glass, phosphors, water treatment and catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57804-25-8
Molecular Formula La2O12S3
Molecular Weight (g/mol) 565.98
MDL Number MFCD00011070
InChI Key VQEHIYWBGOJJDM-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) trisulfate
SMILES [La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point White heat (decomposition)
Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.9% (REO)
Solubility Information Slightly soluble in water.
Formula Weight 566.00 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.821
Chemical Name or Material Lanthanum(III) sulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.