missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) sulfate, anhydrous, 99.9% (REO)

Product Code. 11483820
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11483820

Brand: Thermo Scientific Alfa Aesar 089760.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10099-60-2 | La2O12S3 | 565.979 g/mol

Used for atomic weight determinations for lanthanum. Also used as catalyst and in electronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-60-2
Molecular Formula La2O12S3
Molecular Weight (g/mol) 565.979
MDL Number MFCD00011070
InChI Key VQEHIYWBGOJJDM-UHFFFAOYSA-H
Synonym lanthanum sulfate, sulfuric acid, lanthanum 3+ salt 3:2, dilanthanum 3+ trisulphate, lanthanum iii sulfate 2:3, dilanthanum 3+ trisulfate, lanthansulfat, lanthanum sulphate, lanthanum 3+ trisulfate, 2la.3so4, lanthanum iii sulfatehydrate
PubChem CID 159259
IUPAC Name lanthanum(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[La+3].[La+3]

Specifications

Melting Point 1150°C (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Merck Index 14,5363
Solubility Information Soluble in water.
Formula Weight 566
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.60 g/mL
Assay (REO)
Chemical Name or Material Lanthanum (III) sulfate, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.