missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) phosphate hydrate, 99.99% (REO)

Product Code. p-7051791
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11384987

Brand: Thermo Scientific Alfa Aesar 041990.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14913-14-5 | LaO4P | 233.88 g/mol

It is an active pharmacetical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14913-14-5
Molecular Formula LaO4P
Molecular Weight (g/mol) 233.88
MDL Number MFCD00150248
InChI Key LQFNMFDUAPEJRY-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) phosphate
SMILES [La+3].[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Lump
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 233.88 (Anhydrous)
Percent Purity 99.99%
Odor Odorless
Assay (REO)
Chemical Name or Material Lanthanum(III) phosphate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.