missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) phosphate hydrate, 99.99% (REO)

Product Code. 11384987
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384987 5 g 5g
11374987 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384987 Supplier Thermo Scientific Alfa Aesar Supplier No. 041990.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14913-14-5 | LaO4P | 233.88 g/mol

It is an active pharmacetical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14913-14-5
Molecular Formula LaO4P
Molecular Weight (g/mol) 233.88
MDL Number MFCD00150248
InChI Key LQFNMFDUAPEJRY-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) phosphate
SMILES [La+3].[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Lump
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 233.88 (Anhydrous)
Percent Purity 99.99%
Odor Odorless
Assay (REO)
Chemical Name or Material Lanthanum(III) phosphate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.