missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) perchlorate hexahydrate

Product Code. 11351748
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11351748

Brand: Thermo Scientific Alfa Aesar 011641.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14017-46-0 | Cl3H12LaO18 | 545.33 g/mol

Lanthanum(III) perchlorate hexahydrate participates in the preparation of the Schiff base complexes of lanthanide (III) ions. When hydrated lanthanum perchlorate in ethanolic solution is treated with an ethanolic solution of terpyridine, [La(terpy)3](CIO4)3 is deposited as very small crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14017-46-0
Molecular Formula Cl3H12LaO18
Molecular Weight (g/mol) 545.33
MDL Number MFCD00150245
InChI Key DCEUOMQVLPJQAI-UHFFFAOYSA-K
Synonym lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution
PubChem CID 166977
IUPAC Name lanthanum(3+);triperchlorate
SMILES O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 100 g
UN Number UN1481
Solubility Information Easily soluble in water.
Formula Weight 545.35 (437.26 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Lanthanum(III) perchlorate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.