missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, REacton™, 99.99% (REO)

Product Code. 11306338
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306338 250 g 250g
11396328 50 g 50g
11386328 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11306338 Supplier Thermo Scientific Alfa Aesar Supplier No. 011264.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1312-81-8 | La2O3

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
MDL Number MFCD00011071

Specifications

Boiling Point 4200°C
Melting Point 2307°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water. Soluble in dilute acid.
Formula Weight 325.82
Odor Odorless
Sensitivity Hygroscopic
Density 6.51
Chemical Name or Material Lanthanum(III) oxide, Rare Earth Oxides Content
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.