missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, 99.9%, (trace metal basis)

Product Code. 10410671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10410671 250 g Glass bottle 250g
10398843 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10410671 Supplier Thermo Scientific Acros Supplier No. 199162500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1312-81-8 | La2O3 | 325.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011071

Specifications

Boiling Point 4200.0°C
Melting Point 2315.0°C
Color Blue
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 99.85% min. (La2O3) TREO base
Linear Formula La2O3
Merck Index 15, 5414
Solubility Information Solubility in water: insoluble
Formula Weight 325.82
Percent Purity 99.9%
Grade Trace Metal Basis
Loss on Ignition 3.0 % max. (1000°C)
Packaging Glass bottle
Chemical Name or Material Lanthanum(III) oxide, 99.9%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.