missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacture of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 4200°C |
Melting Point | 2307°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.9% (REO) |
Molecular Formula | La2O3 |
Solubility Information | Insoluble in water but soluble in dilute acid. |
Formula Weight | 325.82 |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'