missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, 91%, Thermo Scientific Chemicals

Catalog Number 10167180
Click to view available options
Quantity:
100 g
250 g
This item is not returnable. View return policy
1312-81-8
La2O3
325.82
MFCD00011071
This item is not returnable. View return policy
1312-81-8
La2O3
325.82
MFCD00011071

Lanthanum(III) oxide, 91%, La2O3, CAS Number-1312-81-8, 100g, 89% min, 4200.0 deg.C, White to Gray, 215-200-5, 325.82, HYGROSCOPIC, GHS H Statement: Causes serious eye irritation.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 4200.0°C
Melting Point 2315.0°C
Color White to Gray
Physical Form Powder
Quantity 250 g
Assay Percent Range 89% min
Linear Formula La2O3
Merck Index 15, 5414
Solubility Information Solubility in water: insoluble.
Formula Weight 325.82
Percent Purity 91%
Packaging Glass bottle
Chemical Name or Material Lanthanum(III) oxide, 91%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'