missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxalate hydrate, 99.9% (REO)

Product Code. 11394788
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394788 500 g 500g
11304798 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394788 Supplier Thermo Scientific Alfa Aesar Supplier No. 043122.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79079-18-8 | C6La2O12 | 541.87 g/mol

Lanthanum Oxalate is mainly applied in specialty glass, phosphors, water treatment and catalyst. This material use in glass, ceramics, paper, textile, coating, fine and specialty chemicals. In petroleum cracking catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79079-18-8
Molecular Formula C6La2O12
Molecular Weight (g/mol) 541.87
MDL Number MFCD00150540
InChI Key OXHNIMPTBAKYRS-UHFFFAOYSA-H
Synonym lanthanum iii oxalate decahydrate, lanthanum oxalate decahydrate, lanthanum iii oxalate, bis 4,5-dioxo-1,3-dioxa-2-lanthanacyclopentan-2-yl oxalate decahydrate
PubChem CID 91886560
IUPAC Name lanthanum;oxalic acid;decahydrate
SMILES [La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 500 g
Linear Formula La2(C2O4)3·xH2O
UN Number UN3288
Solubility Information Soluble in water. Soluble in strong mineral acids.
Formula Weight 541.87 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Lanthanum (III) oxalate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.