missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) nitride, 99.9% (REO)

Product Code. 11348108
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348108 25 g 25g
11358108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348108 Supplier Thermo Scientific Alfa Aesar Supplier No. 039342.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25764-10-7 | LaN | 152.912 g/mol

Lanthanum(III) nitride is used as a precursor along with silicon alkoxide to synthesize La9.33Si6O26, which finds application as electrolytes for solid oxide fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25764-10-7
Molecular Formula LaN
Molecular Weight (g/mol) 152.912
MDL Number MFCD00049962
InChI Key QCLQZCOGUCNIOC-UHFFFAOYSA-N
Synonym lanthanum nitride, lanthanum iii nitride, lanthanumylidyneamine
PubChem CID 117627
IUPAC Name azanylidynelanthanum
SMILES N#[La]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water. Soluble in acid.
Formula Weight 152.91
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Lanthanum(III) nitride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.