missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) nitrate hydrate, 99%, water ca 22-25%, Thermo Scientific™

Product Code. 11452687
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11452687

Brand: Thermo Scientific Alfa Aesar A14890.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Lanthanum(III) nitrate hydrate serves as a raw material for the electrochemical synthesis of LaMnO4, which is a thin film coating on stainless steel substrate. It is also used in preparation of La2O2S. Further, it is used in phosphors, fluorescent display tubes and heat transfer measurement.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100587-94-8
Molecular Formula LaN3O9
Molecular Weight (g/mol) 324.92
MDL Number MFCD00011067
InChI Key FYDKNKUEBJQCCN-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hydrate, lanthanum nitrate hydrate, nitric acid,lanthanum 3+ salt, hydrate 9ci, acmc-1c39v, la.3no3.h2o, lanthanum nitrate-water 1/3/1, lanthanum 3+ hydrate trinitrate, lanthanum 3+ ion hydrate trinitrate, lanthanum nitrate, releasing agent solution, specpure, lanthanum iii nitrate hydrate trace metals basis
PubChem CID 45051674
SMILES [La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 40°C
Quantity 100 g
Linear Formula La(NO3)3·xH2O
UN Number UN1477
Merck Index 14,5363
Solubility Information Soluble in water.
IUPAC Name lanthanum(3+) trinitrate
Formula Weight 324.92 (Anhydrous)
Percent Purity 99%
Assay water ≈22 to 25%
Chemical Name or Material Lanthanum(III) nitrate hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.