missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) isopropoxide, La 40%

Product Code. 11342578
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342578 2 g 2g
11332578 0.5 g 0.5g
11352578 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342578 Supplier Thermo Scientific Alfa Aesar Supplier No. 014616.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19446-52-7 | C9H21LaO3 | 316.169 g/mol

Lanthanum(III) isopropoxide finds application in asymmetric catalysis. It forms complex with an (S,S)-phenyl-linked-binaphthol ,which enhances a catalytic 1,4-addition of malonate to cyclic and acyclic enones. It acts as a co-catalyst with copper fluoride-triphenylphosphine in catalytic allylation of ketoimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19446-52-7
Molecular Formula C9H21LaO3
Molecular Weight (g/mol) 316.169
MDL Number MFCD00015305
InChI Key SORGMJIXNUWMMR-UHFFFAOYSA-N
Synonym lanthanum isopropoxide, lanthanum iii isopropoxide, acmc-20ako6, lanthanum triisopropoxide, lanthanum 3+ ion tris propan-2-olate, lanthanum 3+ tris propan-2-olate
PubChem CID 9858075
IUPAC Name lanthanum(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[La+3]

Specifications

Melting Point 120°C to 128°C
Color White
Boiling Point 170°C (0.04 mmHg)(sublimation)
Odor Alcohol-like
Linear Formula La[OCH(CH3)2]3
Quantity 2 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Soluble in terahydrofuran,toluene,hexane and isopropyl alcohol.
Formula Weight 316.18
Concentration or Composition (by Analyte or Components) La 40%
Physical Form Powder
Chemical Name or Material Lanthanum(III) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.