missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) iodide, ultra dry, 99.995+% (metals basis)

Product Code. 11415310
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415310 1 g 1g
11425310 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415310 Supplier Thermo Scientific Alfa Aesar Supplier No. 044548.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13813-22-4 | I3La | 519.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-22-4
Molecular Formula I3La
Molecular Weight (g/mol) 519.62
MDL Number MFCD00049470
InChI Key KYKBXWMMXCGRBA-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) triiodide
SMILES [I-].[I-].[I-].[La+3]

Specifications

Melting Point 772°C
Physical Form Beads
Quantity 1 g
Formula Weight 519.62
Percent Purity ≥99.995%
Packaging Ampule
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Lanthanum (III) iodide, Ultra dry

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.