missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Product Code. p-4407510
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11386348

Brand: Thermo Scientific Alfa Aesar 011273.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13709-38-1 | F3La | 195.90 g/mol

Lanthanum(III) fluoride is used in phosphor lamp coating, carbon arc electrodes and in lasers. It is also used in fluoride selective electrodes. Further, it is used as a non-ferrous metal additive and used in metal alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-38-1
Molecular Formula F3La
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011073
InChI Key BYMUNNMMXKDFEZ-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trifluoride
SMILES [F-].[F-].[F-].[La+3]

Specifications

Melting Point 1493°C
Color White
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in strong mineral acids. Insoluble in water.
Formula Weight 195.9
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Density 5.936
Chemical Name or Material Lanthanum(III) fluoride, anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.