missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number 11398597
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10099-58-8
Cl3La
245.26
MFCD00011068
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
[Cl-].[Cl-].[Cl-].[La+3]
This item is not returnable. View return policy
10099-58-8
Cl3La
245.26
MFCD00011068
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
[Cl-].[Cl-].[Cl-].[La+3]

Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1,000°C
Melting Point 860°C
Color White
Physical Form Granules
Quantity 100 g
UN Number UN3077
Merck Index 14,5363
Solubility Information Soluble in water.
Formula Weight 245.27
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.842 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'