missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11374788
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
20211-76-1
Cl3La
245.26
MFCD00149753
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
[Cl-].[Cl-].[Cl-].[La+3]
This item is not returnable. View return policy
20211-76-1
Cl3La
245.26
MFCD00149753
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
[Cl-].[Cl-].[Cl-].[La+3]

Lanthanum(III) chloride is useful as precursor for synthesis of aqueous sol-gel lanthanum phosphate nano rods,as a filter aid,an effective flocculent and precipitation phosphate. It is also used as a catalyst for the high pressure oxidative chlorination of methane to chloromethane and organic synthesis it acts as Lewis acid catalyst. It is used as a scintillator material and in Gamma Detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 91°C (decomposition)
pH 5
Physical Form Crystalline
Quantity 100 g
Merck Index 14,5363
Solubility Information Soluble in water and ethanol.
Formula Weight 245.27 (Anhydrous)
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Lanthanum (III) chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'