missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Lanthanum(III) chloride is useful as precursor for synthesis of aqueous sol-gel lanthanum phosphate nano rods,as a filter aid,an effective flocculent and precipitation phosphate. It is also used as a catalyst for the high pressure oxidative chlorination of methane to chloromethane and organic synthesis it acts as Lewis acid catalyst. It is used as a scintillator material and in Gamma Detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 91°C (decomposition) |
pH | 5 |
Physical Form | Crystalline |
Quantity | 500 g |
Merck Index | 14,5363 |
Solubility Information | Soluble in water and ethanol. |
Formula Weight | 245.27 (Anhydrous) |
Percent Purity | 99.9% |
Odor | Odorless |
Assay | (REO) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'