missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate, ACS

Product Code. 11377647
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377647 50 g 50g
11367647 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377647 Supplier Thermo Scientific Alfa Aesar Supplier No. 012916.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 10025-84-0 | Cl3H14LaO7 | 371.36 g/mol

Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]

Specifikationer

Melting Point 91°C (decomposition)
Physical Form Crystalline
Quantity 50 g
Merck Index 14,5363
Solubility Information Soluble in ethanol and water.
Formula Weight 371.37 (245.27 Anhydrous)
Grade ACS
Chemical Name or Material Lanthanum(III) chloride heptahydrate, For analysis ACS
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Lanthanum(III) chloride heptahydrate
Signalord
  • Danger
Riskkategorier
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
Faroangivelser
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Kompletterande information
  • MIXTURE LIST-Contain: Lanthanum(III) chloride heptahydrate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.