missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride, anhydrous, 99.9% (REO)

Product Code. 11339129
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339129 100 g 100g
11329129 5 g 5g
11349129 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339129 Supplier Thermo Scientific Alfa Aesar Supplier No. 087911.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10099-58-8 | Cl3La | 245.26 g/mol

Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Boiling Point 1,000°C
Melting Point 860°C
Color White
Physical Form Powder
Quantity 100 g
UN Number UN3077
Merck Index 14,5363
Solubility Information Soluble in water.
Formula Weight 245.27
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.842 g/mL
Assay (REO)
Chemical Name or Material Lanthanum (III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.