missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) acetate sesquihydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Lanthanum(III) acetate sesquihydrate is used in specialty glass and in water treatment. It is used in the preparation of phenolate-bridged dilanthanum(III) complex, which is considered as a model for metalloproteins in applied chemistry. Further, it is used to prepare nanoparticulate porous lanthanum oxyfluoride (LaOF) films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Lump/Powder |
Quantity | 500 g |
Solubility Information | Soluble in water. |
Formula Weight | 343.05 (316.03 Anhydrous) |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Assay | (REO) |
Chemical Name or Material | Lanthanum(III) acetate sesquihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'