missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) 2-ethylhexanoate, 10% w/v in hexane, Thermo Scientific Chemicals

Product Code. 11369647
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369647 25 mL 25mL
11379647 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369647 Supplier Thermo Scientific Alfa Aesar Supplier No. 041268.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum(III) 2-ethylhexanoate, 10% w/v in hexane is used as drier in ink, coatings and paint industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67816-09-5
Molecular Formula C24H45LaO6
Molecular Weight (g/mol) 568.523
MDL Number MFCD00798555
InChI Key PPNFILUQDVDXDA-UHFFFAOYSA-K
Synonym lanthanum tris 2-ethylhexanoate, lanthanum 2-ethylhexoate, lanthanum 3+ 2-ethylhexanoate, lanthanum 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lanthanum 3+ salt, hexanoic acid, 2-ethyl-, lanthanum 3+ salt 3:1, 2-ethylhexanoate; lanthanum 3+, tris 2-ethylhexanoic acid lanthanum salt, lanthanum 3+ tris 2-ethylhexanoate
PubChem CID 171899
IUPAC Name 2-ethylhexanoate;lanthanum(3+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[La+3]

Specifications

Quantity 25 mL
UN Number UN1208
Formula Weight 568.18
Concentration or Composition (by Analyte or Components) 10% w/v in hexane
Physical Form Liquid
Chemical Name or Material Lanthanum(III) 2-ethylhexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.