Learn More
Lanthanum foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals
£215.00 - £687.00
Chemical Identifiers
CAS | 7439-91-0 |
---|---|
Molecular Formula | La |
Molecular Weight (g/mol) | 138.91 |
MDL Number | MFCD00011066 |
InChI Key | FZLIPJUXYLNCLC-UHFFFAOYSA-N |
Synonym | in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation |
PubChem CID | 23926 |
ChEBI | CHEBI:33336 |
IUPAC Name | lanthanum |
SMILES | [La] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361278
|
Thermo Scientific Alfa Aesar
010220.FF |
25 x 25 mm |
£215.00
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345759
|
Thermo Scientific Alfa Aesar
010220.FI |
50 x 50 mm |
£687.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7439-91-0 | |
138.91 | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
23926 | |
lanthanum |
La | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
CHEBI:33336 | |
[La] |
Specifications
Silver to Gray | |
La | |
UN3208 | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
lanthanum | |
23926 | |
99.9% | |
≈3.9g/25x25mm | |
(REO) |
7439-91-0 | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
[La] | |
138.91 | |
CHEBI:33336 | |
Mylar under Argon | |
Foil | |
Lanthanum foil, 1.0mm (0.04 in.) thick |
Safety and Handling
GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
missing translation for 'einecsNumber' : 231-099-0
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only